Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EDG8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598877
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598877 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598877 Supplier Invitrogen™ Supplier No. PA598877
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EDG8 Polyclonal Antibody for Western Blot, ICC/IF, ELISA

The lysosphingolipid sphingosine 1-phosphate (S1P) regulates cell proliferation, apoptosis, motility, and neurite retraction. Its actions may be both intracellular as a second messenger and extracellular as a receptor ligand. S1P and the structurally related lysolipid mediator lysophosphatidic acid (LPA) signal cells through a set of G protein-coupled receptors known as EDG receptors. Some EDG receptors (e.g., EDG1) are S1P receptors; others (e.g., EDG2) are LPA receptors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EDG8
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene S1PR5
Gene Accession No. Q91X56, Q9H228
Gene Alias Edg8; Edg-8; endothelial differentiation G-protein-coupled receptor 8; endothelial differentiation, sphingolipid G-protein-coupled receptor, 8; lpB4; Lysophospholipid receptor B4; nerve growth factor-regulated G-protein-coupled receptor 1; Nrg1; NRG-1; S1P receptor 5; S1P receptor Edg-8; S1P(5) receptor; S1P5; S1PR5; Sphingosine 1-phosphate receptor 5; sphingosine 1-phosphate receptor EDG8; sphingosine 1-phosphate receptor Edg-8; sphingosine-1-phosphate receptor 5; sphingosine-1-phosphate receptor LPB4; SPPR-1; SPPR-2
Gene Symbols S1PR5
Host Species Rabbit
Immunogen Recombinant Human Sphingosine 1-phosphate receptor 5 protein (309-398AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53637, 94226
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.