Learn More
Ansell Edge™ 48-706 Vend Pack Gloves
Catalog No. 17004283
Click to view available options
Size:
6
7
8
9
10
11
This item is not returnable. View return policy
This item is not returnable. View return policy
Cut and abrasion-resistant glove granting excellent dexterity for medium-duty tasks.
  • Nitrile foam coating: Good grip in dry or slightly oily conditions
  • Enhanced cut protection: Safer handling of sharp edges or materials
  • Breathable and seamless liner: For comfortable and tactile work gloves
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance CE Number, Ecovadis label, EN 388:2016 +A1:2018, EN ISO 21420:2020, Category II, REACH Compliant
Construction Knit
Cuff Material HPPE, Fiber glass, Nylon, Polyester, Spandex
Coating Material Nitrile Foam
Color Gray
Cut Resistance Rating ANSI A3
Hand Specific
Latex Free Yes
Material HPPE, Nylon, Polyester, Spandex, Foam Nitrile
Product Type Cut-resistant Gloves
Size 8
EN388 Standard 4 x 43 C
For Use With (Application) General maintenance, Hardware assembly, Machining / Stamping / Cutting, Molding, Plastic injections, Raw material handling, Seat and frame build
Gender Unisex
Glove Cuff Color Gray
Glove Cuff Style Knit
Liner Material HPPE, Fiber glass, Nylon, Polyester, Spandex
Model 48-706
Puncture Rating ANSI/ISEA 105-16 Level 5
Thumb Style Straight
Yarn Gauge 13
Show More Show Less
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.