Learn More
Ansell Edge™ 48-706 Vend Pack Gloves
Catalog No. 17004284 Shop All Ansell Products
Change view
Click to view available options
Size:
6
7
8
9
10
11
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-004-284 9
17-004-281 6
17-004-282 7
17-004-283 8
17-004-285 10
17-004-286 11
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 17-004-284 Supplier Ansell Supplier No. 48706VP090
Add to Cart
Add to Cart
This item is not returnable. View return policy
Cut and abrasion-resistant glove granting excellent dexterity for medium-duty tasks.
  • Nitrile foam coating: Good grip in dry or slightly oily conditions
  • Enhanced cut protection: Safer handling of sharp edges or materials
  • Breathable and seamless liner: For comfortable and tactile work gloves
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance CE Number, Ecovadis label, EN 388:2016 +A1:2018, EN ISO 21420:2020, Category II, REACH Compliant
Construction Knit
Cuff Material HPPE, Fiber glass, Nylon, Polyester, Spandex
Coating Material Nitrile Foam
Color Gray
Cut Resistance Rating ANSI A3
Hand Specific
Latex Free Yes
Material HPPE, Nylon, Polyester, Spandex, Foam Nitrile
Product Type Cut-resistant Gloves
Size 9
EN388 Standard 4 x 43 C
For Use With (Application) General maintenance, Hardware assembly, Machining / Stamping / Cutting, Molding, Plastic injections, Raw material handling, Seat and frame build
Gender Unisex
Glove Cuff Color Gray
Glove Cuff Style Knit
Liner Material HPPE, Fiber glass, Nylon, Polyester, Spandex
Model 48-706
Puncture Rating ANSI/ISEA 105-16 Level 5
Thumb Style Straight
Yarn Gauge 13