Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell™ EDGE™ 48-707

Catalog No. 17720619
Click to view available options
Size:
6
7
8
9
10
11

Affordable safety gloves, offering superb grip as well as cut and abrasion resistance.

  • Very high cut protection: For robust mechanical protection gloves
  • Good grip and dexterity: Secure handling for precision tasks
  • Anti-static design: Reduced risk of electrostatic shock
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ANSI ABR 5, ANSI Cut A6, Ecovadis label, EN 388:2016 +A1:2018, EN ISO 21420:2020
Coated Area Palm
Construction Knit
Cuff Material Polyester, Fiber glass, Spandex, Nylon, HPPE
Coating Material Polyurethane
Color Gray
Cut Resistance Rating ANSI A6
Hand Specific
Latex Free Yes
Material Polyester
Product Type Mechanical Protection Gloves
Size 7
EN388 Standard EN388:4X43F
For Use With (Application) Aeronautics, Assembly line, Assembling metal parts and components, Handling sharp or irregular metal pieces, Molding, Plastic injections
Gender Unisex
Glove Cuff Color Grey
Glove Cuff Style Knit
Liner Material Polyester, Fiber glass, Spandex, Nylon, HPPE
Lining Knit Lining
Model 48-707
Puncture Rating ANSI/ISEA 105-16 Level 4
Thumb Style Straight
Yarn Gauge 13
Length (English) 8.66 to 10.63 in.
Length (Metric) 220 to 270 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.