Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell™ EDGE™ Collared Coveralls

Catalog No. 19010402 Shop All Ansell Products
Change view
Click to view available options
Size:
Small
Medium
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-010-402 Medium
19-010-403 Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-010-402 Supplier Ansell™ Supplier No. EW20147M
This item is not returnable. View return policy

Ansell™ EDGE™ Collared Coveralls are ideal for workers who rely on consistent quality industrial clothing for protection from workplace risks

  • Applications:
    • Maintaining lab furniture and equipment
    • Packaging and labeling
    • Assembly of parts
    • Equipment maintenance and repair
    • Cleaning of plant and machines
    • Maintenance of plant and machines
    • Inspection
    • Final Assembly
    • Testing
    • Maintenance
TRUSTED_SUSTAINABILITY

Specifications

Product Type Collared Coveralls
Size Medium
Material Microporous Laminate
Color White
Disposable Single-use
Ankle Style Elastic
Cuff Style Elastic
Neck Style Collared
Sterility Non-sterile
Collar Type Standard fold
Latex Free Yes
No. of Pockets None
Quantity 25/Cs.
Style EDGE™ 67-200 Series - Coveralls
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.