Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell EDGE™ 67-050 Series Isolation Gowns

Catalog No. 19156251 Shop All Ansell Products
Change view
Click to view available options
X-Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-156-251 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-156-251 Supplier Ansell Supplier No. EB0521400
This item is not returnable. View return policy

Ansell™ EDGE™ Isolation Gowns are designed for workers who rely on consistent quality industrial clothing for protection from workplace risks.

  • Polyethylene construction
  • Lightweight with high-level liquid protection
  • Thumb loop and elasticated wrists
  • Latex and silicone free

Applications:

  • Maintaining lab furniture and equipment
  • Packaging and labelling
  • Assembly of parts
  • Equipment maintenance and repair
  • Cleaning of plant and machines
  • Maintenance of plant and machines
  • Inspection (picking, checking and marking)
  • Picking parts and accessories
  • Mounting and dismantling
  • Final assembly
  • Production line support & maintenance
  • Testing
  • Maintenance
TRUSTED_SUSTAINABILITY

Specifications

Product Type General Purpose Gown
Size X-Large
Gender Unisex
No. of Pockets None
Material Polyethylene
Color Blue
Disposable Single-use
Sterility Non-sterile
Cuff Style Elastic
Latex Free Yes
Model EDGE™ 67-050 Isolation Gowns
Autoclavable Not Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.