Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EDIL3/DEL1 Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP128632MFV610
Description
EDIL3/DEL1 Polyclonal antibody specifically detects EDIL3/DEL1 in Human, Mouse samples. It is validated for Western Blot, Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
EDIL3/DEL1 | |
Polyclonal | |
Western Blot, Simple Western, Immunocytochemistry/ Immunofluorescence | |
DEL1developmental endothelial locus-1, Developmentally-regulated endothelial cell locus 1 protein, EGF-like repeat and discoidin I-like domain-containing protein 3, EGF-like repeats and discoidin I-like domains 3, Integrin-binding protein DEL1, MGC26287 | |
Synthetic peptide made to an internal portion of human Del-1 (within residues 200-350). [Swiss-Prot# O43854] | |
0.1 mL | |
Angiogenesis, Cell Biology, Cellular Markers, Endothelial Cell Markers, Extracellular Matrix, Signal Transduction | |
10085 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunocytochemistry | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction