Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EDIL3 Mouse anti-Human, Unconjugated, Clone: 4C9, Abnova™
Mouse Monoclonal Antibody.
Supplier: Abnova Corporation H00010085M01J
Description
EDIL3 Monoclonal antibody specifically detects EDIL3 in Human samples. It is validated for WB-Ti, IF, S-ELISA, ELISA, WB-Re.Specifications
EDIL3 | |
Monoclonal | |
Unconjugated | |
EDIL3 | |
DEL1, MGC26287 | |
EDIL3 (NP_005702, 101 a.a. ∼ 199 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa | |
Primary | |
Human | |
IgG2a κ |
Immunofluorescence, ELISA, Western Blot | |
4C9 | |
EGF-like repeats and discoidin I-like domains 3 | |
NM_005711 | |
Mouse | |
100 μg | |
10085 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction