Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EDIL3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597689
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597689 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597689 Supplier Invitrogen™ Supplier No. PA597689
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EDIL3 Polyclonal Antibody for Western Blot, IHC (P), ELISA

Del-1 is an integrin ligand that is expressed by endothelial cells during embryological vascular development. It promotes adhesion of endothelial cells through an interaction with the alpha-v/beta-3 integrin receptor. Del-1 plays a vital role in mediating angiogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EDIL3
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.78 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene EDIL3
Gene Accession No. O43854
Gene Alias Del1; Del-1; developmental endothelial locus-1; developmentally-regulated endothelial cell locus 1 protein; Edil3; EGF like repeats and discoidin domains 3; EGF-like repeat and discoidin I-like domain-containing protein 3; EGF-like repeats and discoidin I-like domains 3; integrin-binding protein DEL1
Gene Symbols EDIL3
Host Species Rabbit
Immunogen Recombinant Human EGF-like repeat and discoidin I-like domain-containing protein 3 protein (60-300AA).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10085
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.