Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDIL3 Rabbit anti-Feline, Human, Mouse, Non-human Primate, Rat, Polyclonal, FabGennix

Catalog No. 502206751
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-6751 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-6751 Supplier Fabgennix Inc Supplier No. DEL101AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EDIL3 Polyclonal antibody specifically detects EDIL3 in Feline, Human, Mouse, Non-human Primate, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen EDIL3
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene EDIL3
Gene Accession No. O35474, O43854
Gene Alias Del1; Del-1; developmental endothelial locus-1; developmentally-regulated endothelial cell locus 1 protein; Edil3; EGF like repeats and discoidin domains 3; EGF-like repeat and discoidin I-like domain-containing protein 3; EGF-like repeats and discoidin I-like domains 3; integrin-binding protein DEL1
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 30-60 on Del-1 protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10085, 101097205, 13612, 471489, 688400
Target Species Feline, Human, Mouse, Non-human Primate, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.