Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ EDRF1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568488
Description
EDRF1 Polyclonal Antibody for Western Blot
This gene may play a role in erythroid cell differentiation. The encoded protein inhibits DNA binding of the erythroid transcription factor GATA-1 and may regulate the expression of alpha-globin and gamma-globin. Alternative splicing results in multiple transcript variants.
Specifications
EDRF1 | |
Polyclonal | |
Unconjugated | |
Edrf1 | |
2700050L05Rik; AU022667; AW558805; C10orf137; EDRF1; erythroid differentiation regulatory factor 1; erythroid differentiation-related factor 1; RGD1306820 | |
Rabbit | |
Affinity chromatography | |
RUO | |
26098 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5-1.0 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q3B7T1 | |
Edrf1 | |
synthetic peptide directed towards the Middle region of human EDRF1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction