Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDTA (Dihydrate), ACS Reagent (Ethylenedinitrilo)tetraacetic Acid, Disodium Salt Dihydrate), Reagents

Catalog No. 01337985
Change view
Click to view available options
Quantity:
125 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-985 500 g
01-337-986 125 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-337-985 Supplier Reagents Holdings Llc Supplier No. C1201100500A5
Add to Cart
Add to Cart

EDTA (Dihydrate), ACS Reagent (Ethylenedinitrilo)tetraacetic Acid, Disodium Salt Dihydrate), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 6381-92-6
Molecular Formula C10H18N2Na2O10
Molecular Weight (g/mol) 372.24
InChI Key OVBJJZOQPCKUOR-UHFFFAOYSA-L
Synonym Edetate Disodium, Disodium EDTA, (Ethylenedinitrilo)tetraacetic Acid disodium salt dihydrate
IUPAC Name disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate
SMILES O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O

Specifications

Melting Point 252°C
Color White
pH 4 to 6
Boiling Point >252°C
Packaging Poly Bottle
Quantity 500 g
Specific Gravity 0.86
Grade ACS
Physical Form Solid
Chemical Name or Material EDTA (Dihydrate), ACS Reagent (Ethylenedinitrilo)tetraacetic Acid, Disodium Salt Dihydrate)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.