Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EDTA (Dihydrate), ACS Reagent (Ethylenedinitrilo)tetraacetic Acid, Disodium Salt Dihydrate), Reagents
Click to view available options
Quantity:
125 g
500 g
10 lb.
10 kg
50 kg
Specifications
Specifications
Melting Point | 252°C |
Color | White |
pH | 4 to 6 |
Boiling Point | >252°C |
Packaging | Poly Pail |
Quantity | 10 kg |
Specific Gravity | 0.86 |
Grade | ACS |
Physical Form | Solid |
Chemical Name or Material | EDTA (Dihydrate), ACS Reagent (Ethylenedinitrilo)tetraacetic Acid, Disodium Salt Dihydrate) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction