Learn More
EDTA, Disodium, 0.100 Molar, Reagents
EDTA, Disodium, 0.100 Molar, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.
Supplier: Reagents Holdings Llc C72011904A
| Quantity | 4 L |
|---|
Chemical Identifiers
| 7732-18-5 , 6381-92-6 | |
| 18.02 | |
| Edetate Disodium, Water Hardness Titrant, (Ethylenedinitrilo)tetraacetic Acid disodium salt | |
| O |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water |
Specifications
| Colorless | |
| 100°C | |
| Edetate Disodium, Water Hardness Titrant, (Ethylenedinitrilo)tetraacetic Acid disodium salt | |
| O | |
| 18.02 | |
| Poly Bottle | |
| 4 to 6 | |
| Non-sterile | |
| EDTA, Disodium, 0.100 Molar |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.