Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EDTA Titrant, 0.100 M with Magnesium, Reagents
EDTA Titrant, 0.100 M with Magnesium, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Reagents Holdings Llc CE2502004A
Specifications
EDTA Titrant, 0.100 M with Magnesium | |
7732-18-5, 6381-92-6 | |
C10H14N2O8Na2/H2O | |
Reagent | |
1.018 | |
4 L | |
Liquid |
Colorless | |
100°C | |
Edetate Disodium, Water Hardness Titrant, (Ethylenedinitrilo)tetraacetic Acid disodium salt | |
Poly Bottle | |
4 to 6 | |
Non-sterile |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction