Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ AIDS Kit II: Simulation of HIV Detection by Western Blot
SDPSCI_ED

Catalog No. s66751
Click to view available options
Class Size:
6 Lab Groups

Includes

instructions, samples, standard molecular weight markers, protein agarose, various buffers and reagents, PVDF membrane, filter paper, stain, 1 ml pipet, 100 ml graduated cylinder

Requires

  • Horizontal gel electrophoresis apparatus
  • Hotplate, micropipets, incubation oven and shaker platform
  • All necessary safety equipment
  • Isopropanol, glacial acetic acid and distilled water

Provides a comprehensive introduction to the molecular biology and pathogenesis of AIDS.

  • Separate protein samples using horizontal agarose electrophoresis
  • Samples are transferred to a membrane and detected by staining
  • Demonstrate the concepts and methodology involved with Western blots
Instructor's Guide provides:
  • Pre-lab preparations and common pitfalls to avoid
  • Answers to study questions and MSDS sheets

Does not contain HIV or its derivatives. No components have been prepared from human sources.

Specifications

Product Type Electrophoresis Kit
Grade 9 to Undergraduate
For Use With (Application) Separate protein samples from hypothetical patients on agarose gels, transfer the samples to a membrane and detect the simulated HIV proteins
Includes Instructions, samples, standard molecular weight markers, protein agarose, various buffers and reagents, PVDF membrane, filter paper, stain, 1 mL pipet, 100 mL graduated cylinder
Quantity 6 Blots
Requires Electrophoresis apparatus, power supply, automatic micropipettes with tips, microwave or hot plate, incubation oven, shaker platform, lab glassware, small plastic trays, microtest tubes, pipet pumps or bulbs, metric rulers, distilled water, methanol, glacial acetic acid
Time Range Electrophoresis: 45 minutes; Blot: overnight; Detection: 25 minutes
Content And Storage Some components require refrigerator storage
Class Size 6 Lab Groups
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.