Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Colored Beads
SDPSCI_ED

Catalog No. S94667 Shop All EDVOTEK Products
Click to view available options
Class Size:
25 Students

Can be designated to represent the Watson-Crick DNA bases (A, T, G, C).

The beads can be used in a variety of ways to demonstrate concepts related to the structure and biology of DNA.

  • With detailed outline of various sample demonstrations
  • 150 of each color

Specifications

Product Type Colored Beads
Grade 6 to 12
For Use With (Application) To demonstrates concepts related to the structure and biology of dna
Includes Detailed outline of various sample demonstrations, 150 beads of each color
Class Size 25 Students
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.