Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Construction and Cloning of DNA Recombinant
SDPSCI_ED

Catalog No. s11301nd Shop All EDVOTEK Products
missing translation for 'orderingAttributeHoverText'
:
For college level courses

Includes

Ultra-pure Water, DNA Fragments for Ligation, T4 DNA Ligase in Reaction Tube, Kanamycin Sulfate, Supercoiled Control DNA Samples, JM109 BactoBeads™, 0.05 M Calcium Chloride, ReadyPour™ LB Agar, Recovery Broth, Growth Medium, Tris-EDTA-Glucose Buffer (TEG), 2M NaOH, 10% SDS, RNase (DNase-free), Tris-EDTA Buffer, Acidified Potassium Acetate, Ultra-pure Water for Restriction Digests, 10X Restriction Enzyme Reaction Buffer, Dryzymes™ Rehydration Buffer, DNA Standard Marker, Supercoiled Plasmid Vector Standard, EcoRI Dryzymes™, PvuII Dryzymes™, XhoI Dryzymes™, InstaStain™ Ethidium Bromide, 10X Gel Loading Solution, 50X Electrophoresis Buffer, UltraSpec-Agarose, Screw Cap Tubes (sterile), Microtest tubes (0.5 ml), petri plates (sterile, 60 x 15mm), 1.5 ml microcentrifuge tubes, sterile inoculating loops/needles, 10ml pipets (sterile), sterile 50 ml culture tubes, petri plates (sterile, 100 x 15 mm)

To study gene structure, function, and to enhance gene expression.

  • It is divided into five modules
  • Clones are constructed by ligation of a vector and a fragment insert
  • The constructs are then transformed into competent cells and the cells are grown and selected for resistance
  • Plasmid DNA is then isolated from the transformants, cleaved with restriction enzymes, and analyzed by agarose gel electrophoresis
  • Recommended for college level courses
  • For 5 lab groups
  • Time: Enzyme digestion 30 min., Gel preparation 30 min., Electrophoresis 45 min., Staining 15 min. and Destaining 20 min.

Unit comprises: Electrophoresis tank and power supply, automatic micropipet with tips, balance, microwave or hot plate, waterbath, large weigh boats for staining, UV transilluminator, floating racks for microtest tubes, pipet pump or bulb, 5 or 10mL pipets, laboratory glassware, metric rulers, distilled water and ice

Specifications

Format Kit
Isolation Technology Tube Centrifugation
For Use With (Equipment) Electrophoresis apparatus
For Use With (Application) To study gene structure, function and to enhance gene expression
Includes Instructions, Enzymes, plasmid DNA, restriction Enzyme dilution Buffer, Enzyme grade Water, standard DNA fragments, restriction Enzyme Reaction Buffer, Gel loading solution, Agarose powder, electrophoresis Buffer, stains, calibrated pipet
Product Type Construction and Cloning Kit
Quantity 5 Groups
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.