Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Detection of a Simulated Infectious Agent
SDPSCI_ED

Catalog No. S67193 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
25 Students
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S67193 25 Students
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S67193 Supplier Edvotek™ Supplier No. 166
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • All necessary materials except those listed under Requires (below)
  • Complete instructions, background information and study questions

Requires

  • Safety goggles
  • Additional long UV Mini-Lights (recommended)
  • Lab coats or protective clothing

Tracking an outbreak by understanding the pattern of disease transmission.

  • Students observe pattern of transmission
  • Dye spots act as surrogates for actual infection
  • Students become 'infected' with dye spots
  • 'Infection' can only be detected under black lights
  • Contains no infectious materials

Order Info

Class size: 10 groups.

Specifications

Grade 9 to Undergraduate
For Use With (Application) Transmit a simulated infectious agent (chemical dye) between classmates
Includes Instructions, reagents for simulating an infectious agent (fluorescent dye indicator and negative sample), test tubes and caps, transfer pipets, one long uv mini light, cotton swabs, petroleum jelly, gloves
Time Range 30 to 45 min.
Class Size 25 Students
Product Type Simulation Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.