Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Edvotek™ Exploring the Infectious Nature of Viruses Experiment Kit

Shop All EDVOTEK Products


Click to view available options
Class Size:
10 Lab Groups
Description
Includes
Instructions, cells, phage beads, Ready-Pour Agar, COLORTOP™ Agar, microtest tubes, sterile loops, petri plates, luria broth, PBS
Requires
37°C incubation oven, waterbath, automatic micropipets with tips
Although bacterial viruses, or bacteriophages, are present in many natural environments, they cannot survive autonomously. They require a host cell to reproduce and survive. In this experiment, students will learn about the different life cycles of bacterial viruses. They will then perform a viral plaque assay to indirectly visualize the viruses and to determine viral titer and multiplicity of infection (MOI).
Specifications
Specifications
| Grade | 9 to Undergraduate |
| Includes | Instructions, cells, phage beads, Ready-Pour Agar, COLORTOP™ Agar, microtest tubes, sterile loops, petri plates, luria broth, PBS |
| Time Range | 2 Hr. |
| Content And Storage | Some components require refrigerator and freezer storage |
| Class Size | 10 Lab Groups |
| Product Type | Exploring the Infectious Nature of Viruses Experiment Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction