Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Genes in a Tube™
SDPSCI_ED

Catalog No. s68635 Shop All EDVOTEK Products
Click to view available options
Class Size:
26 Students

Includes

instructions, lysis buffer, NaCl solution, Protease, Tris buffer, FlashBlue™ solution, microcentrifuge tubes, sterile cotton tipped applicators, transfer pipets, tubes for DNA precipitation, Gene Tubes™, and string

Requires

Ice cold alcohol (95% ethanol or isopropyl alcohol) and a water bath (56°C)

Students can transfer their DNA to a tube that can be worn as a pendant.

  • Students extract and precipitate their own DNA
  • Transfer to tube or save on DNA report card

Specifications

Product Type DNA Report Card Classroom Kit
Grade 6 to 12
For Use With (Application) For extract and spool their own dna in this exciting and easy activity
Includes Lysis buffer, nacl solution, protease, tris buffer, flashblue™ solution, microcentrifuge tubes, sterile cotton tipped applicators, transfer pipets, tubes for dna precipitation, gene tubes™ and string, instructions and s
Requires Ice cold ethanol or isopropanol, waterbath and test tube rack
Time Range 30 min.
Content And Storage Room temperature
Class Size 26 Students
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.