Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Edvotek™ How Do You Clone a Gene?



Click to view available options
Class Size:
5 Lab Groups
Description
Includes
Instructor guide with suggested activities and demonstrations and eight plastic EDVO-links
Use EDVO-links to demonstrate various concepts in biotechnology.
- Plasmids are circular bacterial DNA
- Plasmids act as vehicles that can be genetically engineered to express gene products, such as insulin and growth hormone
- Set of multicolored links demonstrate a variety of molecular biology simulations
- Students learn about digesting DNA with restriction enzymes and cloning genes in plasmids
Order Info
Class size: 10 lab groups.
Specifications
Specifications
Product Type | How Do You Clone A Gene? Experiment |
For Use With (Application) | For learn about digesting dna |
Includes | Instructions, molecular biology models, small plastic bags |
Time Range | 30 min. |
Content And Storage | Room temperature |
Class Size | 5 Lab Groups |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction