Learn More
Edvotek™ Human PCR Tool Box



Description
Includes
Instructions, D1S80 Primer Mix Concentrate, PV92 Primer Mix Concentrate, Mitochondrial Primer Mix Concentrate, EdvoQuick™ DNA Ladder, Control DNA Concentrate, TE Buffer, Proteinase K, UltraSpec-Agarose, Electrophoresis Buffer (50X), 10X Gel Loading Solution, InstaStain™ Ethidium Bromide, FlashBlue™ Stain, Microcentrifuge & PCR Tubes, Disposable Plastic Cups, Salt Packets, & Wax Beads
Requires
Micropipets to measure between 5 and 50μL; Tips; Water bath; Thermal cycler; Electrophoresis apparatus; Power supply; Microwave or hotplate; UV transilluminator
Provides three sets of primers to carry out PCR amplification of:
- Alu element (PV92) on Chromosome 16
- VNTR locus (D1S80) on Chromosome 1
- Two regions of the mitochondrial gene
- Extraction 50 mins
- PCR Setup10 mins
- PCR 2 hours
- Electrophoresis 60 mins
- Staining 5 mins to overnight
Order Info
For 25 Students
Specifications
Specifications
Product Type | PCR Kit |
For Use With (Application) | Provides three sets of primers to carry out the pcr amplification of alu element (pv92) on chromosome 16, the vntr locus (d1s80) on chromosome 1 and two regions of the mitochondrial gene |
Includes | Instructions; proteinase k; pcr edvobeads™; control and primer dna; microtubes; TE buffer; agarose; dna ladder; practice gel loading solution; gel loading dye; electrophoresis buffer; instastain™ ethidium bromide |
Time Range | 2 Hr. |
Content And Storage | Refrigerated |
Class Size | 25 Students |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.