Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Identification of Bacterial Protein Profiles
SDPSCI_ED

Catalog No. s05649 Shop All EDVOTEK Products
Click to view available options
:
Bacterial Protein Identification Kit
This item is not returnable. View return policy

Catalog No. S05649

Supplier: Edvotek™ 252

This item is not returnable. View return policy

Includes

  • Instructions
  • Bacterial cultures and reagents
  • LyphoProteins™
  • Lysozyme, buffers
  • Protein InstaStain™
  • Practice gel loading solution
  • Protein sample buffer
  • Unknown proteins ready for electrophoresis
  • ReadyPour™ Agar
  • Nutrient broth

Compare total protein extracts from several bacterial sources.

  • The unique patterns of protein bands, obtained by SDS vertical polyacrylamide electrophoresis, can be used to identify various bacterial strains

Experiment times:

  • Day one setup, 15 minutes
  • Day two setup, one hour
  • Electrophoresis, one hour
  • Staining, 20 minutes
  • Destaining, two hours

All You Need: 

  • 3 polyacrylamide gels (12%)
  • Vertical gel electrophoresis apparatus
  • Power supply
  • Microcentrifuge
  • Incubation oven
  • Hot plate or burner
  • White light visualization system
  • Automatic micropipet with fine tips
  • Lab glassware
  • Methanol
  • Glacial acetic acid
  • Distilled water.

Order Info

For six groups sharing three gels.

Recommended Storage

Some components require freezer storage

Specifications

For Use With (Application) Compare total protein extracts from several bacterial sources
Class Size 6 Lab Groups
Product Type Fingerprinting of Bacterial Proteins
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.