Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ InstaStain™ Ethidium Bromide
SDPSCI_ED

Catalog No. S05699 Shop All EDVOTEK Products
Click to view available options
Quantity:
7 x 7 cm, for 100 Gels
7 x 7 cm, for 40 Gels

Rapid and sensitive.

  • Contains only a few micrograms of ethidium bromide
  • Gel is ready for visualization in two minutes
  • Much less ethidium bromide waste for disposal compared to standard staining procedure

Specifications

Color Dark Red/Purple
Format Strips
Label or Dye Ethidium bromide
Product Type Fluorescent DNA/RNA Stain
Stain Type Ethidium Bromide
Form Sheet, Solid
For Use With (Application) Rapid and sensitive, disposal of InstaStain™ Ethidium Bromide is minimal compared to the volume of liquid waste generated from the standard Ethidium Bromide staining procedure
Quantity 7 x 7 cm, for 100 Gels
Sufficient For 100 Gels
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.