Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Edvotek™ BactoBeads™ Lyophilized Bacterial Cultures and Cells

Shop All EDVOTEK Products


Click to view available options
Quantity:
5 Beads
Description
E. coli host for transformation with pUC plasmids
Specifications
Specifications
| Content And Storage | Store in freezer. |
| Form | Lyophilized |
| For Use With (Application) | Transformation with pUC plasmids |
| Product Type | Bacterial Culture |
| Quantity | 5 Beads |
| Source | E. coli JM109 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction