Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Principles of PCR
SDPSCI_ED

Catalog No. s11103nd Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
8 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S11103ND 8 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S11103ND Supplier Edvotek™ Supplier No. 103
Only null left
Add to Cart
Add to Cart

Includes

Instructions, Ready-to-Load QuickStrip™ DNA Samples, UltraSpec-Agarose™, Electrophoresis Buffer (50X), 10X Gel Loading Solution, FlashBlue™ DNA Stain, InstaStain™ Blue Cards, & Disposable Pipets.

Requires

Horizontal electrophoresis apparatus; Staining tray; Water bath (65°C); Visualization system (white light); Micropipets; 250mL beakers; Hot hand protector; Deionized water; Microwave or hotplate; Balance; Photodocumentation system (optional)

Uses gel electrophoresis to gain an understanding of the Polymerase Chain Reaction (PCR) process and its applications.

  • Simulation experiment does not require a thermal cycler
  • Does not contain human DNA

Specifications

Grade 9 to Undergraduate
Quantity 8 Gels
Class Size 8 Lab Groups
Product Type Electrophoresis Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.