Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Purification and Size Determination of Green and Blue Fluorescent Proteins
SDPSCI_ED

Catalog No. s05651 Shop All EDVOTEK Products
Click to view available options
Class Size:
6 Lab Groups

Includes

  • Instructions
  • Columns and matrix
  • GFP and BFP extracts
  • Buffer
  • Protein gel reagents for optional activity

The unique fluorescent properties of GFP and BFP will be used as an assay during their purification from an E. coli extract.

When bacteria are used to make medicinally useful proteins by transformation, the protein of interest must be separated from all of the other cellular proteins.

  • Unique fluorescent properties of GFP and BFP are used as an assay during their purification from an E. coli extract
  • Column fractions containing GFP or BFP are identified by fluorescence and then purified
  • As an optional activity, purified protein fractions can be separated by SDS polyacrylamide gel electrophoresis to estimate the purity and size of the GFP and BFP proteins

Experiment times:

  • Packing and running column, 45 minutes
  • Optional electrophoresis, one hour
  • Staining, 30 minutes
  • Destaining, two hours

All You Need

  • water bath
  • long wave UV lamp
  • ring stand; clamps
  • automatic micropipet
  • vertical gel electrophoresis apparatus
  • power supply
  • hot plate
  • polyacrylamide gels (12%)

Order Info

For six lab groups.

Recommended Storage

Some components require freezer storage

Specifications

Grade 9 to Undergraduate
For Use With (Application) Used as an assay during their purification from an e. coli extract
Includes Instructions, columns and matrix, gfp and bfp extracts, buffer, protein gel reagents for optional activity
Time Range 4 hours 15 minutes; Staining time: 30 minutes
Content And Storage Refrigerated. Some components require freezer storage.
Class Size 6 Lab Groups
Product Type Green and Blue Fluorescent Protein Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.