Learn More
Description
Includes
Instructions, PCR EdvoBeads™, LyphoPrimer™ Mix, EdvoQuick™ DNA Ladder, LyphoTemplate™ Lambda DNA, TE Buffer, UltraSpec-Agarose™, Electrophoresis Buffer (50X), 10X Gel Loading Solution, InstaStain™ Ethidium Bromide, FlashBlue™ Stain, Microcentrifuge & PCR Tubes, & Wax Beads
- Experiment uses a set of primers and lambda DNA template in a time-saving two-step process
- Allows lab to be completed in one session
- Does not contain human DNA.
Order Info
Class size: 10 lab groups.
Specifications
Specifications
| Grade | 9 to Undergraduate |
| For Use With (Application) | Uses pcr to amplify a small section of lambda dna via a two-step process, which saves valuable classroom time and allows for completion of the lab in one session |
| Includes | Instructions; pcr edvobeads™; DNA template and primers; dna size ladder; ultrapure water; wax beads; gel loading dye; agarose; electrophoresis buffer; instastain™ ethidium bromide and flashblue™ stain; TE buffer |
| Time Range | 60 min. |
| Content And Storage | Refrigerated; Store at: -20°C; -4°F |
| Class Size | 10 Lab Groups |
| Product Type | Quick PCR Classroom Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

