Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Rainbow Transformation Kit
SDPSCI_ED

Catalog No. s21224
Click to view available options
Class Size:
10 Lab Groups

Includes

Instructions, BactoBeads™, plasmid DNA, IPTG, ampicillin, transformation solution, ReadyPour™ Agar, Luria broth, petri dishes, sterile pipets, loops and microtubes

Requires

Waterbath, 37°C incubation oven, microwave or hot plate

Allows for the selection, propagation, expression and purification of a gene.

Students transform bacteria with a new set of rainbow color plasmids that transform non-pathogenic bacterial cells into bright, colorful cells

  • For 10 Lab Groups
  • Set Up and Plating - 50 minutes
  • Overnight incubation
  • Transformation efficiency - 15 minutes

Specifications

Grade 9 to Undergraduate
Includes Instructions, BactoBeads™, plasmid DNA, IPTG, ampicillin, transformation solution, ReadyPour™ Agar, Luria broth, petri dishes, sterile pipets, loops and microtubes
Content And Storage Some components require refrigerator and freezer storage
Class Size 10 Lab Groups
Product Type Rainbow Transformation Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.