Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Reagents for Electrophoresis: Gel-Loading Solution
SDPSCI_ED

Catalog No. s11606 Shop All EDVOTEK Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S11606 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S11606 Supplier Edvotek™ Supplier No. 606
Only null left
Add to Cart
Add to Cart

Includes

Yields 5 ml final volume of DNA sample

Used for electrophoresis.

This concentrated solution is used to prepare nucleic acid and protein samples for agarose or polyacrylamide gel electrophoresis. Gel loading solution includes protein denaturants and the negatively charged tracking dye, Bromophenol Blue. Recommended Usage: Add 1 volume solution to 9 volumes of sample and mix well

Specifications

Color Dark Blue
Concentration 10X
Format Tube
Label or Dye Bromophenol Blue
Form Liquid
For Use With (Application) Agarose or Polyacrylamide Gel electrophoresis
Quantity 1 mL
Product Type Gel-loading Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.