Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Separation of RNA and DNA by Gel Filtration Chromatography
SDPSCI_ED

Catalog No. s05639 Shop All EDVOTEK Products
Change view
Click to view available options
:
Gel Filtration Chromatography
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S05639 Gel Filtration Chromatography
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S05639 Supplier Edvotek™ Supplier No. 204
This item is not returnable. View return policy

Includes

  • Instructions
  • DNA/RNA LyphoSample™
  • Dry matrix
  • Elution buffer
  • Chromatography columns
  • Agarose
  • Gel loading solution
  • Buffer
  • InstaStain™ Blue and FlashBlue™ stain

Requires

Electrophoresis tank; Power supply; Variable micropipet and tips; Balance ; Microwave or hotplate

Purified fractions of DNA and RNA are analyzed by agarose gel electrophoresis.

  • Gel filtration chromatography separates molecules on the basis of size and shape
  • Provides a LyphoSample™ mixture of RNA and DNA that is separated on a gel exclusion column
  • Complete in one hour and 30 minutes

Order Info

Contains materials for five separations and five gels.

Specifications

Grade 9 to Undergraduate
For Use With (Application) Separates Molecules on The Basis of Size and Shape
Includes Instructions; DNA or RNA lyphosample; dry matrix; elution buffer; chromatography columns; agarose; gel loading solution; buffer; instastain blue and flashblue stain
Class Size 5 Lab Groups
Product Type Gel Filtration Chromatography Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.