Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Edvotek™ In Search of the Invisible DNA: A Genetics Exploration

Shop All EDVOTEK Products


Click to view available options
Class Size:
10 Lab Groups
Description
Includes
Instructions, Ready-to-Load QuickStrip™ Dye Samples, UltraSpec-Agarose™, Electrophoresis Buffer (50X), Practice Gel Loading Solution, Disposable Pipets
Requires
Electrophoresis tank; Power supply; Black light
In this lesson, we explore how DNA technology can be used to explore the relationship between genotype and phenotype using one of two exciting scenarios (medical diagnostics or alien genetics). Fluorescent dyes simulate DNA fragments, eliminating post-electrophoresis staining and saving you valuable classroom time!
Specifications
Specifications
Product Type | Classroom Kit |
For Use With (Application) | To perform electrophoresis of dye samples that only become visible when excited by uv light |
Time Range | 45 min. |
Content And Storage | Room temperature |
Class Size | 10 Lab Groups |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction