Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Water Quality Testing I: Chromogenic Analysis of Water Bacteria Contaminants
SDPSCI_ED

Catalog No. s68636 Shop All EDVOTEK Products
Click to view available options
Class Size:
10 Lab Groups

Includes

instructions, BactoBeads™, ReadyPour™ Agar, fluorescent reagents, Petri dishes, inoculating loops, sterile swabs, microtubes

Requires

Longwave UV lamp, microscope, slides and coverslips

Students will test for coliforms in simulated contaminated water, and can test water samples from the environment.

Safe drinking water is vitally important to health. Both pathogenic and harmless bacteria can be found in the guts of mammals and birds. Presence of coliforms in water is an indicator of fecal contamination.

  • Testing for coliforms, including E. coli, using color and fluorescent reagents
  • Extension activity includes Gram Stain test that can be performed on the collected samples
  • Test duration: complete in 30 minutes and grow overnight

Specifications

Grade 9 to Undergraduate
Includes Instructions, BactoBeads™, coliform detection broth, indole detection reagent, nutrient broth, sterile water, tubes, sterile pipets, and inoculating loops
Requires Long wave UV lamp
Time Range Complete in 30 minutes and grow overnight
Content And Storage Some components require refrigerator storage.
Class Size 10 Lab Groups
Product Type Water Test Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.