Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Water Quality Testing III: Multiplexed PCR for Testing Water Bacteria
SDPSCI_ED

Catalog No. s95132 Shop All EDVOTEK Products
Click to view available options
:
Water Quality Testing II
This item is not returnable. View return policy
This item is not returnable. View return policy

Includes

instructions, PCR EdvoBeads™, Universal DNA Buffer, LyphoPrimer™ Mix, EdvoQuick™ DNA Ladder, LyphoControl™ (Complete PCR Control), TE Buffer, Proteinase K, Potassium Acetate, DNA Extraction Buffer, BactoBeads™, UltraSpec-Agarose™, Electrophoresis Buffer (50X), SYBR™ Safe Stain, FlashBlue™ Stain, Microcentrifuge & PCR Tubes

Requires

Thermal cycler; waterbath, electrophoresis tank; power supply; micropipets to measure between 5 and 50μL; tips and microcentrifuge

Students will investigate presence of contaminants in a water sample.

  • Drinking water is routinely tested for contamination. If a screening tests positive, more sophisticated tests are required to determine whether harmful microbes are present
  • Students use PCR in multiplex format
  • Single PCR test detects presence of three separate, classroom-safe organisms
  • Analyzed on agarose gels

Experiment times:

  • Setup: 60 minutes
  • PCR: 120 minutes or overnight
  • Electrophoresis: 45 minutes

Specifications

For Use With (Application) To investigate presence of contaminants in a water sample
For Use With (Equipment) Multiplex PCR
Sufficient For 25 Students
Type Water Quality Testing II
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.