Learn More
Edwards Fittings for BOC High Vacuum Pumps: Centering Ring
For use with BOC high vacuum pumps
Specifications
For Use With BOC high vacuum pump
View More Specs
Products 3
Catalog Number Mfr. No. Material Type Price Quantity & Availability  
Catalog Number Mfr. No. Material Type Price Quantity & Availability  
11-283-31
View Documents
Edwards
C10514395
Polymer NW25/25 Viton Centering Ring N/A N/A N/A
11-282-30
View Documents
Edwards
C10512395
Polymer NW16 Viton Centering Ring N/A N/A N/A
11-283-32
View Documents
Edwards
C10516395
Stainless Steel NW40 Centering Ring N/A N/A N/A
Description
Description
  • Aluminum centering rings manufactured from high-tech polymer
  • Unique slotted design prevents gas bursts
  • CX2 polymer can be used at temperatures up to 120°C and is unaffected by most common solvents
  • Outgassing rate of the material is 6.6 × 10-8mbar l s-1cm-2
Specifications
Specifications
BOC high vacuum pump
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.