Learn More
Edwards Fittings for BOC High Vacuum Pumps: Swing Clamp
Shop All Edwards High Vacuum International Products
Click to view available options
Type:
NW10/16 Swing clamp
NW20/25 Swing Clamp
NW32/40 Swing Clamp
Description
- Compact stainless-steel clamping ring
- Lightweight polymer clamp
- Rugged pneurop standard aluminum clamp
- Metal clamp suitable for aluminum and indium seals wide temperature range
- Claw clamps and bolts provide wide range of applications
Specifications
Specifications
For Use With | BOC high vacuum pump |
Material | Polymer |
Size | 90 x 66 x 35 mm |
Type | NW32/40 Swing Clamp |