Learn More
Edwards Fittings for BOC High Vacuum Pumps: T-Piece
For use with BOC high vacuum pumps
Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
11-283-25
|
Edwards
C10514411 |
NW25 T-Piece | N/A | N/A | N/A | ||||
Description
- Made of aluminum
- Aluminum is ideal for achieving dependable cost-effective performance down to 10-7mbar
C10516395, C10512395, C10514395
- Aluminum centering rings manufactured from high-tech polymer
- Unique slotted design prevents gas bursts
- CX2 polymer can be used at temperatures up to 120°C and is unaffected by most common solvents
- Outgassing rate of the material is 6.6 × 10-8mbar l s-1cm-2
- Nitrile «O« rings:
- Low outgassing rate
- High deformation resistance
- High resistance to most oils
- Fluoroelastomer:
- Very low outgassing rate especially after baking
- Chemical stability at high temperatures up to 200°C
- High resistance to oils and many chemicals
- Compact stainless-steel clamping ring
- Lightweight polymer clamp
- Rugged pneurop standard aluminum clamp
- Metal clamp suitable for aluminum and indium seals wide temperature range
- Claw clamps and bolts provide wide range of applications
Specifications
Aluminum |