Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Edwardsiella Tarda Antibody (236/128), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10062540AF405
Description
Edwardsiella Tarda Monoclonal specifically detects Edwardsiella Tarda in Bacteria samples. It is validated for ELISA.Specifications
Edwardsiella Tarda | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Mouse | |
Protein G purified | |
RUO | |
This product recognizes Edwardsiella tarda. E. tarda is a member of the family Enterobactericeae associated with freshwater ecosystems that that can cause gastoenteritis, wound infections and death due to septicaemia. It is thought that the pathogenicity of this bacteria is due to it's ability to resist phagocytic killing by the host defences. Studies show that replication of E. tarda in murine macrophages is dependant on the type III secretion system and induces an anti-apoptotic effect by up-regulating anti-apoptotic NF-kB target genes. | |
Store at 4C in the dark. | |
IgG1 |
ELISA | |
236/128 | |
ELISA | |
aracolobactrum anguillimortiferum, Edwardsiella anguillimortifera | |
Native | |
0.1 mL | |
Primary | |
Bacteria | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction