Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EEA1 Antibody (6D4.1D4), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP236568AF405
Description
EEA1 Monoclonal specifically detects EEA1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Flow (Intracellular).Specifications
EEA1 | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
early endosome antigen 1, early endosome antigen 1, 162kD, early endosome-associated protein, Endosome-associated protein p162, MSTP105, ZFYVE2, ZFYVE2MST105, Zinc finger FYVE domain-containing protein 2 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Flow Cytometry | |
6D4.1D4 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Flow (Intracellular) | |
Q15075 | |
EEA1 | |
Partial recombinant human EEA1 protein (between amino acids 50-300). [UniProt Q15075] | |
0.1 mL | |
Endosome Markers, Membrane Trafficking and Chaperones | |
8411 | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction