Learn More
EEA1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301898AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
Binds phospholipid vesicles containing phosphatidylinositol 3-phosphate and participates in endosomal trafficking.Specifications
EEA1 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q15075, Q8BL66 | |
Eea1 | |
Between 1361 and 1411 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
EEA1 | |
EEA1, Endosome-associated protein p162, Zinc finger FYVE domain-containing protein 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
216238, 8411 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.