Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EEA1, Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89035701 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-701 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-701 Supplier Abnova Corporation Supplier No. PAB30208

Rabbit Polyclonal Antibody

Specifications

Antigen EEA1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5-10 ug/mL) Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS, pH 7.4 (0.02% sodium azide).
Gene early endosome antigen 1
Gene Alias MST105/MSTP105/ZFYVE2
Gene Symbols EEA1
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human EEA1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8411
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C on dry atmosphere, lyophilized antibodies are stable. After reconstitution with deionized water, store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.