Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EED Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298765PE
Description
EED Polyclonal antibody specifically detects EED in Human samples. It is validated for Western BlotSpecifications
EED | |
Polyclonal | |
Western Blot | |
embryonic ectoderm development, HEED, polycomb protein EED, WAIT1, WAIT-1, WD protein associating with integrin cytoplasmic tails 1 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human EED | |
0.1 mL | |
DNA replication Transcription Translation and Splicing | |
8726 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
PE | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction