Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EED Monoclonal Antibody (41D), Active Motif™

Mouse Monoclonal Antibody
$156.65 - $638.65
Specifications
Antigen | EED |
---|---|
Clone | 41D |
Concentration | 1 mg/mL |
Applications | ChIP Assay, ChIP sequencing (ChIP-seq), Immunohistochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-3233
![]() ![]() |
Active Motif
61204 |
10 μg |
Each for $156.65
|
|
|||||
50-199-3232
![]() ![]() |
Active Motif
61203 |
100 μg |
Each for $638.65
|
|
|||||
Description
The EED protein (WAIT-1 or WD protein associated with integrin cytoplasmic tails-1), also called embryonic ectoderm development, is a member of the superfamily of WD-40 repeat proteins and widely conserved polycomb group (PcG) family of proteins. The polycomb group (PcG) is a large and evolutionarily conserved set of genes whose products act in multimeric complexes to modify histones, which are then thought to cause stable and heritable states of transcriptional repression. EED shuttles between the nucleus and the plasma membrane and can interact with the cytoplasmic tail of integrin Beta-7 subunit. EED exerted an antiviral activity at the late stage of HIV-1 replication.Specifications
EED | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 30% glycerol and 0.035% sodium azide; pH 7.5 | |
EED; embryonic ectoderm development; Embryonic ectoderm development protein; ENSMUSG00000039373; hEED; l(7)5Rn; l7Rn5; lethal, Chr 7, Rinchik 5; lusk; Polycomb protein EED; WAIT1; WAIT-1; WD protein associating with integrin cytoplasmic tails 1 | |
EED | |
IgG2a | |
Protein A | |
Antibody |
41D | |
ChIP Assay, ChIP sequencing (ChIP-seq), Immunohistochemistry | |
Unconjugated | |
Mouse | |
Human | |
O75530 | |
8726 | |
This EED antibody was raised against full-length recombinant human EED protein. | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
EED |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title