Learn More
Invitrogen™ EED Polyclonal Antibody

Description
EED Polyclonal Antibody for Western Blot, IHC (P), ELISA
Specifications
Specifications
| Antigen | EED |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.35 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | EED |
| Gene Accession No. | O75530 |
| Gene Alias | EED; embryonic ectoderm development; Embryonic ectoderm development protein; ENSMUSG00000039373; HEED; l(7)5Rn; l7Rn5; lethal, Chr 7, Rinchik 5; lusk; Polycomb protein EED; WAIT1; WAIT-1; WD protein associating with integrin cytoplasmic tails 1 |
| Gene Symbols | EED |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.