Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EED Recombinant Mouse Monoclonal Antibody, AbFlex™, Active Motif™
SDP

Catalog No. 501993852
Encompass
Change view
Click to view available options
Quantity:
10 μg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-3852 10 μg
50-199-3851 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-3852 Supplier Active Motif Supplier No. 91136
Add to Cart
Add to Cart

Mouse Recombinant Monoclonal Antibody

The EED protein (WAIT-1 or WD protein associated with integrin cytoplasmic tails-1), also called embryonic ectoderm development, is a member of the superfamily of WD-40 repeat proteins and widely conserved polycomb group (PcG) family of proteins. The polycomb group (PcG) is a large and evolutionarily conserved set of genes whose products act in multimeric complexes to modify histones, which are then thought to cause stable and heritable states of transcriptional repression. EED shuttles between the nucleus and the plasma membrane and can interact with the cytoplasmic tail of integrin Beta-7 subunit. EED exerted an antiviral activity at the late stage of HIV-1 replication.

Specifications

Antigen EED
Applications ChIP sequencing (ChIP-seq), ELISA
Classification Monoclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 200mM HEPES, 50mM sodium acetate with 100mM NaCl, 30% glycerol and 0.035% sodium azide; pH 7.5
Gene EED
Gene Accession No. O75530
Gene Alias EED; embryonic ectoderm development; Embryonic ectoderm development protein; ENSMUSG00000039373; hEED; l(7)5Rn; l7Rn5; lethal, Chr 7, Rinchik 5; lusk; Polycomb protein EED; WAIT1; WAIT-1; WD protein associating with integrin cytoplasmic tails 1
Gene Symbols EED
Host Species Mouse
Immunogen This EED antibody was raised against full-length recombinant human EED protein.
Purification Method Metal-chelate chromatography
Quantity 10 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8726
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.