Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
eEF1A1 Mouse anti-Human, PE, Clone: 23C11, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259411PE
This item is not returnable.
View return policy
Description
eEF1A1 Monoclonal antibody specifically detects eEF1A1 in Human samples. It is validated for Flow CytometrySpecifications
eEF1A1 | |
Monoclonal | |
PE | |
CCS3, CCS-3, cervical cancer suppressor 3, CTCL tumor antigen, EE1A1, EEF-1, EEF1A, eEF1A-1, EF1AHNGC:16303, EF1a-like protein, EF-1-alpha-1, EF-Tu, elongation factor 1 alpha subunit, elongation factor 1-alpha 1, Elongation factor Tu, Eukaryotic elongation factor 1 A-1, eukaryotic translation elongation factor 1 alpha 1, eukaryotic translation elongation factor 1 alpha 1-like 14, FLJ25721, glucocorticoid receptor AF-1 specific elongation factor, GRAF-1EF, LENG7, leukocyte receptor cluster (LRC) member 7, Leukocyte receptor cluster member 7, MGC102687, MGC131894, MGC16224, prostate tumor-inducing protein 1, PTI1, translation elongation factor 1 alpha 1-like 14 | |
Recombinant human eEF1A1 (1-462aa) purified from E.coli | |
0.1 mL | |
DNA replication Transcription Translation and Splicing | |
1915 | |
Store at 4°C in the dark. | |
IgG2a κ |
Flow Cytometry | |
23C11 | |
PBS | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction