Learn More
EEF1A1 Mouse anti-Human, Unconjugated, Clone: 1A9F5, Proteintech
Mouse Monoclonal Antibody
Supplier: Proteintech Group Inc 674951IG150UL
Description
The EEF1A1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human EEF1A1. This antibody recognizes human, mouse, rat antigen. The EEF1A1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
EEF1A1 | |
Monoclonal | |
500 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence | |
1A9F5 | |
Unconjugated | |
150 μL | |
1915 | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.