Learn More
EEF1A1 Polyclonal Antibody, Rockland™

Chicken Polyclonal Antibody
Supplier: Rockland Immunochemicals 200901MP6S

Description
Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This affinity purified antibody is directed against human EEF1A1. This product is an IgY fraction antibody purified from monospecific chicken egg yolks by a multi-step process which includes selective precipitation and salt fractionation followed by extensive dialysis against the buffer stated above. A BLAST analysis was used to suggest cross-reactivity with the antigen based on 100% homology with the immunizing sequence to EEF1A1 human, mouse, rat, bovine, rabbit, cat, horse, Chinese hamster, Gallus gallus, Pan troglodytes, and Pongoabelii. May have cross-reactivity to EEF1A to Danio rerio; and eef1as to Xenopuslaevis.
This protein promotes the GTP-dependent binding of aminoacyl-tRNA to the A-site of ribosomes during protein biosynthesis. With PARP1 and TXK, forms a complex that acts as a T helper 1 (Th1) cell-specific transcription factor and binds the promoter of IFN-gamma to directly regulate its transcription, and is thus involved importantly in Th1 cytokine production.Specifications
EEF1A1 | |
Polyclonal | |
Unconjugated | |
Eef1a1 | |
CCS3; CCS-3; cervical cancer suppressor 3; CTCL tumor antigen; EE1A1; EEF-1; EEF1A; Eef1a1; eEF1A-1; eef1a1.S; Eef1a2; Eef1a2l1; eef1as; eef1a-s; EF-1 alpha; EF-1 alpha2; EF1A; EF1A1; EF1a-like protein; EF1alpha; EF-1-alpha-1; EF-1-alpha-S; EF-Tu; elongation factor 1 A-1; elongation factor 1 alpha subunit; Elongation factor 1-alpha 1; Elongation factor 1-alpha, somatic form; Elongation factor Tu; elongation factor-1 alpha; Eukaryotic elongation factor 1 A-1; eukaryotic translation elongation factor 1 alpha 1; eukaryotic translation elongation factor 1 alpha 1 S homeolog; eukaryotic translation elongation factor 1 alpha 1-like 14; eukaryotic translation elongation factor 1 alpha 2 like 1; eukaryotic translation elongation factor 1 alpha, somatic form; glucocorticoid receptor AF-1 specific elongation factor; GRAF 1EF; GRAF-1EF; HNGC:16303; LENG7; leukocyte receptor cluster (LRC) member 7; Leukocyte receptor cluster member 7; prostate tumor-inducing protein 1; PTI1; SI; translation elongation factor 1 alpha 1-like 14; XELAEV_18029462mg | |
Chicken | |
25 μL | |
Primary | |
Human | |
Antibody |
ELISA, Immunohistochemistry, Western Blot | |
0.95 mg/mL | |
0.002M sodium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7 | |
P68104 | |
Eef1a1 | |
Anti-EEF1A1 antibody was prepared from eggs of chickens laid after repeated immunizations with a synthetic peptide corresponding to a N-terminal portion of human EEF1A1 conjugated to Keyhole Limpet Hemocyanin (KLH). | |
RUO | |
1915 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.