Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
eEF1A1 Rabbit anti-Human, DyLight 594, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299359DL594
Description
eEF1A1 Polyclonal specifically detects eEF1A1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
eEF1A1 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
DyLight 594 | |
CCS3, CCS-3, cervical cancer suppressor 3, CTCL tumor antigen, EE1A1, EEF-1, EEF1A, eEF1A-1, EF1AHNGC:16303, EF1a-like protein, EF-1-alpha-1, EF-Tu, elongation factor 1 alpha subunit, elongation factor 1-alpha 1, Elongation factor Tu, Eukaryotic elongation factor 1 A-1, eukaryotic translation elongation factor 1 alpha 1, eukaryotic translation elongation factor 1 alpha 1-like 14, FLJ25721, glucocorticoid receptor AF-1 specific elongation factor, GRAF-1EF, LENG7, leukocyte receptor cluster (LRC) member 7, Leukocyte receptor cluster member 7, MGC102687, MGC131894, MGC16224, prostate tumor-inducing protein 1, PTI1, translation elongation factor 1 alpha 1-like 14 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human eEF1A1. | |
0.1 mL | |
DNA replication Transcription Translation and Splicing | |
1915 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction